Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allyl (3-Methylbutoxy)acetate 98.0+%, TCI America™
SDP

Catalog No. A31855G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A31855G 5 g
A318525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A31855G Supplier TCI America Supplier No. A31855G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 67634-00-8
Molecular Formula C10H18O3
Molecular Weight (g/mol) 186.251
MDL Number MFCD03424212
InChI Key XCWPXUNHSPOFGV-UHFFFAOYSA-N
Synonym (3-Methylbutoxy)acetic Acid Allyl Ester
PubChem CID 106729
IUPAC Name prop-2-enyl 2-(3-methylbutoxy)acetate
SMILES CC(C)CCOCC(=O)OCC=C

Specifications

Color Yellow
Quantity 5 g
Formula Weight 186.25
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Allyl (3-Methylbutoxy)acetate
RTECSNumber AI8988000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.