accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNH3CONOS(E/Z)

CAS RN 677331-12-3

IUPAC Name: 5-{[2,5-dimethyl-1-(pyridin-3-yl)-1H-pyrrol-3-yl]methylidene}-3-phenyl-1,3-thiazolidine-2,4-dione
Synonyms: 5-{[2,5-dimethyl-1-(pyridin-3-yl)pyrrol-3-yl]methylidene}-3-phenyl-1,3-thiazolidine-2,4-dione
Molecular Weight (g/mol): 375.45
Molecular Formula: C21H17N3O2S
InChi Key: NCSHZXNGQYSKLR-UHFFFAOYSA-N
SMILES: CC1=CC(C=C2SC(=O)N(C2=O)C2=CC=CC=C2)=C(C)N1C1=CN=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)