accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OH3COCH3(E)

CAS RN 67746-30-9

IUPAC Name: (2E)-1,1-diethoxyhex-2-ene
Synonyms: (2E)-1,1-diethoxyhex-2-ene
Molecular Weight (g/mol): 172.27
Molecular Formula: C10H20O2
InChi Key: WMQKYHTZGYIHHD-CMDGGOBGSA-N
SMILES: CCC\C=C\C(OCC)OCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results