Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-2-Hexen-1-al Diethyl Acetal 95.0+%, TCI America™
SDP

Catalog No. H057525ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H057525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H057525ML Supplier TCI America Supplier No. H057525ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 67746-30-9
Molecular Formula C10H20O2
Molecular Weight (g/mol) 172.268
MDL Number MFCD00009472
InChI Key WMQKYHTZGYIHHD-CMDGGOBGSA-N
Synonym 1,1-Diethoxy-trans-2-hexene, Leaf Aldehyde Diethyl Acetal
PubChem CID 5365137
IUPAC Name (E)-1,1-diethoxyhex-2-ene
SMILES CCCC=CC(OCC)OCC

Specifications

Color Yellow
Boiling Point 98°C
Quantity 25 mL
Formula Weight 172.27
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material trans-2-Hexen-1-al Diethyl Acetal
EINECSNumber (2)-0516
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.