accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 67769-47-5

IUPAC Name: dilithium(1+) 8-amino-3-[(hydrazinecarbonyl)amino]-2,4-dioxo-3-azatricyclo[7.3.1.0⁵,¹³]trideca-1(13),5,7,9,11-pentaene-7,11-disulfonate
Synonyms: lucifer yellow carbohydrazide dye
Molecular Weight (g/mol): 457.24
Molecular Formula: C13H9Li2N5O9S2
InChi Key: RPKCZJYDUKVMGF-UHFFFAOYSA-L
SMILES: [Li+].[Li+].NNC(=O)NN1C(=O)C2=CC(=C(N)C3=CC(=CC(C1=O)=C23)S([O-])(=O)=O)S([O-])(=O)=O
Color 
  • (5)
  • (3)
Molecular Weight (g/mol) 
  • (5)
  • (3)
Quantity 
  • (2)
  • (2)
  • (3)
  • (1)
Percent Purity 
  • (2)
  • (3)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (5)
  • (3)
  • (5)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (3)
  • (3)