Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lucifer yellow CH, Dilithium salt, ≥99%, >95%, MP Biomedicals

Catalog No. ICN15526725 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15526725 25 mg
ICN15526710 10 mg
ICN15526780 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15526725 Supplier MP Biomedicals, Inc Supplier No. 0215526725
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fluorescent dye used for staining nerve cells and demonstrating electrical coupling.

  • Chemical Name: Lucifer yellow CH, Dilithium salt
  • Physical Form: Powder
  • Purity: ≥99%
  • Molecular Formula: C13H9Li2N5O9S2
  • Molecular Weight: 457.241 g/mol
  • UV/Vis Absorbance: 426 ± 5 nm (max in water: 427 nm)
  • Key Applications: Fluorescent labels, fluorescent probes, and stains
  • Use Cases: Staining nerve cells; demonstrating electrical coupling between hamster oocytes and cumulous cells
  • Caution: For research use only
  • Storage: 2°C to 8°C, store under nitrogen, and desiccate
  • Assay: >95%
  • CAS number: 67769-47-5
  • Storage conditions: 2°C to 8°C, desiccate, store under nitrogen
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67769-47-5
Molecular Formula C13H9Li2N5O9S2
Molecular Weight (g/mol) 457.241
InChI Key RPKCZJYDUKVMGF-UHFFFAOYSA-L
Synonym 6-amino-2[(Hyrazinocarbonyl)amino]-2,3-dihydro-1,3-dioxo-1H-Benz[de] Isoquinoline-5,8-disulfonic acid dilithium salt
PubChem CID 124220108
ChEBI CHEBI:52106
IUPAC Name dilithium;6-amino-2-(hydrazinecarbonylamino)-1,3-dioxobenzo[de]isoquinoline-5,8-disulfonate
SMILES [Li+].[Li+].C1=C(C=C2C3=C1C(=O)N(C(=O)C3=CC(=C2N)S(=O)(=O)[O-])NC(=O)NN)S(=O)(=O)[O-]

Specifications

Assay Percent Range >95%
Color Orange to Yellow
Quantity 25 mg
Physical Form Powder
Formula Weight 457.2
Percent Purity ≥99%
Grade Research
Chemical Name or Material Lucifer yellow CH, Dilithium salt, For HPLC analysis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.