accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6777-00-0

IUPAC Name: 3-(prop-2-en-1-yloxy)oxetane
Synonyms: 3-(prop-2-en-1-yloxy)oxetane
Molecular Weight (g/mol): 114.14
Molecular Formula: C6H10O2
InChi Key: NONFBYUIHZQHQT-UHFFFAOYSA-N
SMILES: C=CCOC1COC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results