accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3BrSBrBr

CAS RN 67869-13-0

IUPAC Name: 2,3,5-tribromo-4-methylthiophene
Synonyms: 2,3,5-tribromo-4-methylthiophene
Molecular Weight (g/mol): 334.85
Molecular Formula: C5H3Br3S
InChi Key: DTLNXOZXOZCNMX-UHFFFAOYSA-N
SMILES: CC1=C(Br)SC(Br)=C1Br
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results