accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C(R)H3CCH3(R)NH3(R)NH3

CAS RN 67907-32-8

IUPAC Name: (1R,2R,5R)-2-(azaniumylmethyl)-3,3,5-trimethylcyclopentan-1-aminium
Synonyms: (1R,2R,5R)-2-(ammoniomethyl)-3,3,5-trimethylcyclopentan-1-aminium
Molecular Weight (g/mol): 158.29
Molecular Formula: C9H22N2
InChi Key: QKHWUKPTSMULMZ-BWZBUEFSSA-P
SMILES: C[C@@H]1CC(C)(C)[C@H](C[NH3+])[C@@H]1[NH3+]
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results