accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 67963-68-2

IUPAC Name: (4-bromophenoxy)(tert-butyl)dimethylsilane
Synonyms: 4-bromophenoxy(tert-butyl)dimethylsilane
Molecular Weight (g/mol): 287.27
Molecular Formula: C12H19BrOSi
InChi Key: DLGZGLKSNRKLSM-UHFFFAOYSA-N
SMILES: CC(C)(C)[Si](C)(C)OC1=CC=C(Br)C=C1
Boiling Point 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results