accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 67969-82-8

IUPAC Name: boron(3+) hydrogen ethoxyethane tetrafluoride
Synonyms: boron(3+) hydrogen diethyl ether tetrafluoride
Molecular Weight (g/mol): 161.93
Molecular Formula: C4H11BF4O
InChi Key: GYFCYMJGLCBLMX-UHFFFAOYSA-K
SMILES: [H+].[B+3].[F-].[F-].[F-].[F-].CCOCC
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)