accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HNH3COOH3CNHONHNOCH3OOCH3CH3OOHHO

CAS RN 679839-66-8

IUPAC Name: 2-hydroxypropanoic acid; 3,5-dimethyl 4-{3-[({3-[4-(3-methoxyphenyl)piperidin-1-yl]propyl}carbamoyl)amino]phenyl}-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
Synonyms: 3,5-dimethyl 4-{3-[({3-[4-(3-methoxyphenyl)piperidin-1-yl]propyl}carbamoyl)amino]phenyl}-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate; lactic acid
Molecular Weight (g/mol): 680.8
Molecular Formula: C36H48N4O9
InChi Key: BYHJIPSVXAFCDI-UHFFFAOYNA-N
SMILES: CC(O)C(O)=O.COC(=O)C1=C(C)NC(C)=C(C1C1=CC(NC(=O)NCCCN2CCC(CC2)C2=CC(OC)=CC=C2)=CC=C1)C(=O)OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)