accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOCH3O(Z)

CAS RN 68133-76-6

IUPAC Name: (3Z)-hex-3-en-1-yl 2-oxopropanoate
Synonyms: (3Z)-hex-3-en-1-yl 2-oxopropanoate
Molecular Weight (g/mol): 170.21
Molecular Formula: C9H14O3
InChi Key: LKNXTZXOBHAYSR-PLNGDYQASA-N
SMILES: CC\C=C/CCOC(=O)C(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results