accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 682-09-7

IUPAC Name: 2,2-bis[(prop-2-en-1-yloxy)methyl]butan-1-ol
Synonyms: 2,2-bis[(prop-2-en-1-yloxy)methyl]butan-1-ol
Molecular Weight (g/mol): 214.31
Molecular Formula: C12H22O3
InChi Key: BDKDHWOPFRTWPP-UHFFFAOYSA-N
SMILES: CCC(CO)(COCC=C)COCC=C
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)