accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH3(R)OOCl

CAS RN 68208-21-9

IUPAC Name: (3R)-3-azaniumyl-3-(3-chlorophenyl)propanoate
Synonyms: (3R)-3-ammonio-3-(3-chlorophenyl)propanoate
Molecular Weight (g/mol): 199.63
Molecular Formula: C9H10ClNO2
InChi Key: LIDRHPCWOYOBIZ-MRVPVSSYSA-N
SMILES: [NH3+][C@H](CC([O-])=O)C1=CC=CC(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results