Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-3-(3-chlorophenyl)propionic acid, 98%

Catalog No. AAL1979703
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1979703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1979703 Supplier Thermo Scientific Chemicals Supplier No. L1979703
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 68208-21-9 | C9H10ClNO2 | 199.634 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68208-21-9
Molecular Formula C9H10ClNO2
Molecular Weight (g/mol) 199.634
MDL Number MFCD00463160
InChI Key LIDRHPCWOYOBIZ-UHFFFAOYSA-N
Synonym 3-amino-3-3-chlorophenyl propanoic acid, 3-amino-3-3-chloro-phenyl-propionic acid, dl-beta-3-chlorophenyl alanine, 3-amino-3-3-chlorophenyl propionic acid, 3-3-chlorophenyl-dl-beta-alanine, 3-3-chlorophenyl-beta-alanine, dl-3-amino-3-3-chloro-phenyl-propionic acid, r-3-3-chlorophenyl-beta-alanine, rarechem ak hc t320, dl--3-chlorophenyl alanine
PubChem CID 2764178
IUPAC Name 3-amino-3-(3-chlorophenyl)propanoic acid
SMILES C1=CC(=CC(=C1)Cl)C(CC(=O)O)N

Specifications

Melting Point ∽217°C (decomposition)
Beilstein 2938807
Quantity 1 g
Formula Weight 199.64
Percent Purity 98%
Chemical Name or Material 3-Amino-3-(3-chlorophenyl)propionic acid
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.