accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3H3CH3CH3CNCH3H3CH3CCNNCNN

CAS RN 68271-98-7

IUPAC Name: bis(tetrabutylazanium); {2-[4'-(2-azanidylene-1-cyanoeth-1-en-1-yl)-[1,1'-biphenyl]-4-yl]-2-cyanoeth-1-en-1-ylidene}azanide
Synonyms: bis(tetrabutylammonium ion); {2-[4'-(2-azanidylene-1-cyanoeth-1-en-1-yl)-[1,1'-biphenyl]-4-yl]-2-cyanoeth-1-en-1-ylidene}azanide
Molecular Weight (g/mol): 765.23
Molecular Formula: C50H80N6
InChi Key: NJFQTNXYFOEFJU-UHFFFAOYSA-N
SMILES: CCCC[N+](CCCC)(CCCC)CCCC.CCCC[N+](CCCC)(CCCC)CCCC.[N-]=C=C(C#N)C1=CC=C(C=C1)C1=CC=C(C=C1)C(C#N)=C=[N-]
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results