accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 68278-23-9

IUPAC Name: hydrogen 2,5-diamino-2-(difluoromethyl)pentanoic acid chloride
Synonyms: hydrogen vaniqa chloride
Molecular Weight (g/mol): 218.63
Molecular Formula: C6H13ClF2N2O2
InChi Key: VKDGNNYJFSHYKD-UHFFFAOYNA-N
SMILES: [H+].[Cl-].NCCCC(N)(C(F)F)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results