accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOF

CAS RN 682784-98-1

IUPAC Name: N-benzyl-2-fluoro-N-phenylbenzamide
Synonyms: N-benzyl-2-fluoro-N-phenylbenzamide
Molecular Weight (g/mol): 305.35
Molecular Formula: C20H16FNO
InChi Key: DNHHEHCSAPIQBC-UHFFFAOYSA-N
SMILES: FC1=CC=CC=C1C(=O)N(CC1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results