accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOHO

CAS RN 6836-22-2

IUPAC Name: 2-(7-methoxynaphthalen-1-yl)acetic acid
Synonyms: (7-methoxynaphthalen-1-yl)acetic acid
Molecular Weight (g/mol): 216.24
Molecular Formula: C13H12O3
InChi Key: GJPYHKXILUFWKV-UHFFFAOYSA-N
SMILES: COC1=CC=C2C=CC=C(CC(O)=O)C2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results