Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Methoxy-1-naphthaleneacetic Acid 98.0+%, TCI America™
SDP

Catalog No. M28885G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M28885G 5 g
M28881G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M28885G Supplier TCI America Supplier No. M28885G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6836-22-2
Molecular Formula C13H12O3
Molecular Weight (g/mol) 216.236
MDL Number MFCD12756806
InChI Key GJPYHKXILUFWKV-UHFFFAOYSA-N
Synonym (7-Methoxy-1-naphthyl)acetic Acid
PubChem CID 13570105
IUPAC Name 2-(7-methoxynaphthalen-1-yl)acetic acid
SMILES COC1=CC2=C(C=CC=C2CC(=O)O)C=C1

Specifications

Melting Point 157°C
Color White-Yellow
Quantity 5 g
Formula Weight 216.24
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 7-Methoxy-1-naphthaleneacetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.