accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOCH3CH3OOH3CH3C

CAS RN 684238-37-7

IUPAC Name: 3,3,6,6-tetramethyl-9-{4-[(2-methylphenyl)methoxy]phenyl}-2,3,4,5,6,7,8,9-octahydro-1H-xanthene-1,8-dione
Synonyms: 3,3,6,6-tetramethyl-9-{4-[(2-methylphenyl)methoxy]phenyl}-4,5,7,9-tetrahydro-2H-xanthene-1,8-dione
Molecular Weight (g/mol): 470.61
Molecular Formula: C31H34O4
InChi Key: UEKIYVKPQNKSDI-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1COC1=CC=C(C=C1)C1C2=C(CC(C)(C)CC2=O)OC2=C1C(=O)CC(C)(C)C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)