accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNHOOHOHOO

CAS RN 684283-16-7

IUPAC Name: 1-(phenanthrene-2-carbonyl)piperazine-2,3-dicarboxylic acid
Synonyms: 1-(phenanthrene-2-carbonyl)piperazine-2,3-dicarboxylic acid
Molecular Weight (g/mol): 378.38
Molecular Formula: C21H18N2O5
InChi Key: IWWXIZOMXGOTPP-UHFFFAOYNA-N
SMILES: OC(=O)C1NCCN(C1C(O)=O)C(=O)C1=CC=C2C(C=CC3=CC=CC=C23)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results