accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3(E)

CAS RN 68480-29-5

IUPAC Name: (3E)-hex-3-en-1-yl non-2-ynoate
Synonyms: (3E)-hex-3-en-1-yl non-2-ynoate
Molecular Weight (g/mol): 236.36
Molecular Formula: C15H24O2
InChi Key: PDPNRGJWKDIKKR-SOFGYWHQSA-N
SMILES: CCCCCCC#CC(=O)OCC\C=C\CC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results