accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6851-80-5

IUPAC Name: [(2-methoxyphenyl)methyl](methyl)azanium
Synonyms: [(2-methoxyphenyl)methyl](methyl)azanium
Molecular Weight (g/mol): 152.22
Molecular Formula: C9H14NO
InChi Key: JCCQJCOMFAJJCQ-UHFFFAOYSA-O
SMILES: C[NH2+]CC1=CC=CC=C1OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results