accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OSOONHNOHN

CAS RN 685506-42-7

IUPAC Name: 3-[2-methyl-5-(piperidine-1-sulfonyl)furan-3-yl]-1-phenylurea
Synonyms: 3-[2-methyl-5-(piperidine-1-sulfonyl)furan-3-yl]-1-phenylurea
Molecular Weight (g/mol): 363.43
Molecular Formula: C17H21N3O4S
InChi Key: LVOVQRPAMXCXTM-UHFFFAOYSA-N
SMILES: CC1=C(NC(=O)NC2=CC=CC=C2)C=C(O1)S(=O)(=O)N1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results