accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OCH3CH3

CAS RN 6863-58-7

IUPAC Name: 2-(butan-2-yloxy)butane
Synonyms: DI (sec-butyl) ether diisobutylene oxide
Molecular Weight (g/mol): 130.23
Molecular Formula: C8H18O
InChi Key: HHBZZTKMMLDNDN-UHFFFAOYNA-N
SMILES: CCC(C)OC(C)CC
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)