Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

sec-Butyl Ether (DL- and meso- mixture) (stabilized with HQ) 97.0+%, TCI America™
SDP

Catalog No. D14055ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D14055ML 5 mL
D140525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D14055ML Supplier TCI America Supplier No. D14055ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6863-58-7
Molecular Formula C8H18O
Molecular Weight (g/mol) 130.231
MDL Number MFCD00039921
InChI Key HHBZZTKMMLDNDN-UHFFFAOYSA-N
Synonym Di-sec-butyl Ether
PubChem CID 23294
IUPAC Name 2-butan-2-yloxybutane
SMILES CCC(C)OC(C)CC

Specifications

Color Colorless
Boiling Point 121°C
Quantity 5 mL
UN Number 1149
Formula Weight 130.23
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material sec-Butyl Ether (DL- and meso- mixture) (stabilized with HQ)
EINECSNumber (2)-0363
RTECSNumber EQ2220000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.