accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOOHOSiCH3H3CCH3CH3H3CH3C

CAS RN 68703-51-5

IUPAC Name: 6-{[(tert-butyldimethylsilyl)oxy]methyl}-2,2-dimethyl-tetrahydro-2H-furo[3,4-d][1,3]dioxol-4-ol
Synonyms: 6-{[(tert-butyldimethylsilyl)oxy]methyl}-2,2-dimethyl-tetrahydrofuro[3,4-d][1,3]dioxol-4-ol
Molecular Weight (g/mol): 304.46
Molecular Formula: C14H28O5Si
InChi Key: JJLRGSYXWAKGJM-UHFFFAOYNA-N
SMILES: CC(C)(C)[Si](C)(C)OCC1OC(O)C2OC(C)(C)OC12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results