accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2OH2ONaNaNaNaOPOOOOOOPOOO

CAS RN 68807-90-9

IUPAC Name: tetrasodium dihydrate 4-{3-oxo-1-[4-(phosphonatooxy)phenyl]-1,3-dihydro-2-benzofuran-1-yl}phenyl phosphate
Synonyms: tetrasodium dihydrate 4-{3-oxo-1-[4-(phosphonatooxy)phenyl]-2-benzofuran-1-yl}phenyl phosphate
Molecular Weight (g/mol): 602.24
Molecular Formula: C20H16Na4O12P2
InChi Key: QYFYAAPDTZOYPC-UHFFFAOYSA-J
SMILES: O.O.[Na+].[Na+].[Na+].[Na+].[O-]P([O-])(=O)OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(OP([O-])([O-])=O)C=C1
Color 
  • (1)
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results