accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHSOON

CAS RN 68922-18-9

IUPAC Name: 2-(pyridin-2-yl)ethane-1-sulfonic acid
Synonyms: 2-(pyridin-2-yl)ethanesulfonic acid
Molecular Weight (g/mol): 187.21
Molecular Formula: C7H9NO3S
InChi Key: VRBVUAYEXFCDPE-UHFFFAOYSA-N
SMILES: OS(=O)(=O)CCC1=CC=CC=N1
Grade 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results