accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClH3CNCH3NSCl

CAS RN 69-09-0

IUPAC Name: hydrogen [3-(2-chloro-10H-phenothiazin-10-yl)propyl]dimethylamine chloride
Synonyms: chlorpromazine hydrochloride chlorpromazinium chloride hebanil hibernal plegomazin chlorazin marazine ampliactil aminazin propaphenin
Molecular Weight (g/mol): 355.32
Molecular Formula: C17H20Cl2N2S
InChi Key: FBSMERQALIEGJT-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CN(C)CCCN1C2=CC=CC=C2SC2=CC=C(Cl)C=C12
Molecular Weight (g/mol) 
  • (10)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (3)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (2)
  • (1)
Grade 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (5)
  • (1)
  • (2)
  • (3)
  • (10)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)