accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaCH3CH3S(R)(R)NHO(R)NH2ON(S)OO

CAS RN 69-52-3

IUPAC Name: sodium (2S,5R,6R)-6-[(2R)-2-amino-2-phenylacetamido]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
Synonyms: ampicillin sodium ampicillin natrium binotal sodium ampicillin austrapen
Molecular Weight (g/mol): 371.39
Molecular Formula: C16H18N3NaO4S
InChi Key: KLOHDWPABZXLGI-YWUHCJSESA-M
SMILES: [Na+].CC1(C)S[C@@H]2[C@H](NC(=O)[C@H](N)C3=CC=CC=C3)C(=O)N2[C@H]1C([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (3)
  • (5)
Molecular Weight (g/mol) 
  • (5)
  • (5)
Physical Form 
  • (3)
  • (4)
  • (2)
Type 
  • (1)
  • (7)
Grade 
  • (1)
  • (1)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (3)
  • (5)
  • (5)
  • (5)
  • (3)
  • (4)
  • (2)
  • (1)
  • (7)
  • (1)
  • (1)
  • (1)
  • (3)
  • (5)
  • (1)
  • (3)
  • (1)