Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ampicillin Sodium Salt, G-Biosciences
SCI_ED

Catalog No. S157239
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S157239 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S157239 Supplier G-Biosciences Supplier No. RC852
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ampicillin is an inhibitor of bacterial cell wall synthesis which acts by inhibiting the cross-linking of peptidoglycan.

It is used in the selection of antibiotic cell hybrids, and has a high rate of action against many gram negative bacterial strains.

Chemical Identifiers

CAS 69-52-3

Specifications

Type α-Carboxybenzyl Penicillin Disodium Salt
Physical Form Powder
Sterility Non-sterile
Color White
Quantity 1 kg
Percent Purity >95%
Grade Molecular Biology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.