Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ampicillin sodium salt, 94.9%, MP Biomedicals™

Catalog No. ICN19014880 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19014880 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19014880 Supplier MP Biomedicals, Inc Supplier No. 0219014880
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used to select for ampicillin resistance in mutated and transformed cells

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69-52-3

Specifications

Color White
Form Powder
Purity or Quality Grade 94.9%
Quantity 100 g
Chemical Name or Material Ampicillin sodium salt
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.