CAS RN 69011-18-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Ambertec™ UP550 OH Anion Exchange Resin, Sold by MilliporeSigma™ Supelco™
Dowex™ Monosphere™ 550A UPW OH type 1 strong base anion exchange resin is recommended for the roughing, intermediate, and polishing ion exchange applications. Low TOC levels in UPW grade are achieved by deliberate functionalization and rinsing.