accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CNHNCH3

CAS RN 690632-22-5

IUPAC Name: 1-[(2,4,6-trimethylphenyl)methyl]-1,4-diazepane
Synonyms: 1-[(2,4,6-trimethylphenyl)methyl]-1,4-diazepane
Molecular Weight (g/mol): 232.37
Molecular Formula: C15H24N2
InChi Key: FNKQURSLYXVLGQ-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(CN2CCCNCC2)C(C)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results