Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(Mesitylmethyl)-1,4-diazepane, 97%, Thermo Scientific™

Catalog No. AW00753EA
Change view
Click to view available options
Quantity:
10 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AW00753EA 10 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AW00753EA Supplier Thermo Scientific Chemicals Supplier No. AW00753EA

Chemical Identifiers

CAS 690632-22-5
Molecular Formula C15H24N2
Molecular Weight (g/mol) 232.371
InChI Key FNKQURSLYXVLGQ-UHFFFAOYSA-N
Synonym 1-mesitylmethyl-1,4-diazepane, 1-2,4,6-trimethylphenyl methyl-1,4-diazepane, 1-2,4,6-trimethylbenzyl-1,4-diazepane, 1-2,4,6-trimethylphenyl methyl-1,4-diazaperhydroepine
PubChem CID 2794562
IUPAC Name 1-[(2,4,6-trimethylphenyl)methyl]-1,4-diazepane
SMILES CC1=CC(=C(C(=C1)C)CN2CCCNCC2)C

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 232.37
Chemical Name or Material 1-(Mesitylmethyl)-1, 4-diazepane
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.