accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 690632-36-1

IUPAC Name: 2-(pyrrolidin-1-yl)pyridine-3-carboxylic acid
Synonyms: 2-(pyrrolidin-1-yl)pyridine-3-carboxylic acid
Molecular Weight (g/mol): 192.22
Molecular Formula: C10H12N2O2
InChi Key: DTUSJCJZQJAPGD-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(N=CC=C1)N1CCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results