accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3NOO

CAS RN 693-57-2

IUPAC Name: 12-azaniumyldodecanoate
Synonyms: 12-ammoniododecanoate
Molecular Weight (g/mol): 215.34
Molecular Formula: C12H25NO2
InChi Key: PBLZLIFKVPJDCO-UHFFFAOYSA-N
SMILES: [NH3+]CCCCCCCCCCCC([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results