CAS RN 6933-49-9
IUPAC Name:
2-methoxy-9H-carbazole
Synonyms:
2-methoxy-9H-carbazole
Molecular Weight (g/mol):
197.24
Molecular Formula:
C13H11NO
InChi Key:
MDKVPSJRUXOFFV-UHFFFAOYSA-N
SMILES:
COC1=CC2=C(C=C1)C1=C(N2)C=CC=C1
1
–
1
of
1
results
1
2-Methoxycarbazole 98.0+%, TCI America™
CAS: 6933-49-9 Molecular Formula: C13H11NO Molecular Weight (g/mol): 197.237 MDL Number: MFCD09033506 InChI Key: MDKVPSJRUXOFFV-UHFFFAOYSA-N PubChem CID: 10797986 IUPAC Name: 2-methoxy-9H-carbazole SMILES: COC1=CC2=C(C=C1)C3=CC=CC=C3N2