accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrOO

CAS RN 6940-49-4

IUPAC Name: 3-bromo-1,3-dihydro-2-benzofuran-1-one
Synonyms: 3-bromo-3H-2-benzofuran-1-one
Molecular Weight (g/mol): 213.03
Molecular Formula: C8H5BrO2
InChi Key: CLMSHAWYULIVFQ-UHFFFAOYNA-N
SMILES: BrC1OC(=O)C2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results