accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NH3CN

CAS RN 69478-77-9

IUPAC Name: 1-benzyl-N,N-dimethylpyrrolidin-3-amine
Synonyms: 1-benzyl-N,N-dimethylpyrrolidin-3-amine
Molecular Weight (g/mol): 204.32
Molecular Formula: C13H20N2
InChi Key: SOTUMZCQANFRAM-UHFFFAOYNA-N
SMILES: CN(C)C1CCN(CC2=CC=CC=C2)C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results