Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Benzyl-3-(dimethylamino)pyrrolidine 97.0+%, TCI America™
SDP

Catalog No. B15595G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B15595G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B15595G Supplier TCI America Supplier No. B15595G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 69478-77-9
Molecular Formula C13H20N2
Molecular Weight (g/mol) 204.32
MDL Number MFCD00191304
InChI Key SOTUMZCQANFRAM-UHFFFAOYNA-N
PubChem CID 10013168
IUPAC Name 1-benzyl-N,N-dimethylpyrrolidin-3-amine
SMILES CN(C)C1CCN(CC2=CC=CC=C2)C1

Specifications

Color Yellow
Boiling Point 117°C
Quantity 5 g
UN Number 2735
Formula Weight 204.32
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-Benzyl-3-(dimethylamino)pyrrolidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.