accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6948-86-3

CH3ONCH3OH3COCH3OCH3

CAS RN 6948-86-3

IUPAC Name: bis(2,2-diethoxyethyl)(methyl)amine
Synonyms: bis(2,2-diethoxyethyl)(methyl)amine
Molecular Weight (g/mol): 263.38
Molecular Formula: C13H29NO4
InChi Key: DPLOXPRJCFWURN-UHFFFAOYSA-N
SMILES: CCOC(CN(C)CC(OCC)OCC)OCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results