accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 69573-75-7

IUPAC Name: 1,1,1-trichloro-2-(chloromethoxy)ethane
Synonyms: 1,1,1-trichloro-2-(chloromethoxy)ethane
Molecular Weight (g/mol): 197.86
Molecular Formula: C3H4Cl4O
InChi Key: GXHPCVJGAHVTGO-UHFFFAOYSA-N
SMILES: ClCOCC(Cl)(Cl)Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results