Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloromethyl 2,2,2-Trichloroethyl Ether 97.0+%, TCI America™
SDP

Catalog No. C24125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C24125G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C24125G Supplier TCI America Supplier No. C24125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 69573-75-7
Molecular Formula C3H4Cl4O
Molecular Weight (g/mol) 197.86
MDL Number MFCD20257906
InChI Key GXHPCVJGAHVTGO-UHFFFAOYSA-N
PubChem CID 11019830
IUPAC Name 1,1,1-trichloro-2-(chloromethoxy)ethane
SMILES ClCOCC(Cl)(Cl)Cl

Specifications

Color Yellow
Boiling Point 84°C
Quantity 5 g
UN Number 2922
Formula Weight 197.86
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Chloromethyl 2,2,2-Trichloroethyl Ether
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.