accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6959-71-3

IUPAC Name: 3-butoxypropanenitrile
Synonyms: propanenitrile, 3-butoxy-
Molecular Weight (g/mol): 127.19
Molecular Formula: C7H13NO
InChi Key: AWVNJBFNHGQUQU-UHFFFAOYSA-N
SMILES: CCCCOCCC#N
Boiling Point 
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)