Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Butoxypropionitrile 96.0+%, TCI America™
SDP

Catalog No. B095925ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B095925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B095925ML Supplier TCI America Supplier No. B095925ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6959-71-3
Molecular Formula C7H13NO
Molecular Weight (g/mol) 127.19
MDL Number MFCD00045643
InChI Key AWVNJBFNHGQUQU-UHFFFAOYSA-N
Synonym 3-butoxypropionitrile, propanenitrile, 3-butoxy, beta-butoxy propionitrile, propionitrile, 3-butoxy, 3-butoxypropannitril, 3-butoxypropiononitrile, 3-n-butoxypropionitrile, 3-butoxypropannitril czech, .beta.-butoxy propionitrile, 3-butoxypropanitrile
PubChem CID 23395
IUPAC Name 3-butoxypropanenitrile
SMILES CCCCOCCC#N

Specifications

Color Yellow
Boiling Point 206°C
Quantity 25 mL
Formula Weight 127.19
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Butoxypropionitrile
EINECSNumber (2)-1543
RTECSNumber UG1225000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.