accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 696-23-1

IUPAC Name: 2-methyl-5-nitro-1H-imidazole
Synonyms: 2-methyl-5-nitroimidazole
Molecular Weight (g/mol): 127.1
Molecular Formula: C4H5N3O2
InChi Key: FFYTTYVSDVWNMY-UHFFFAOYSA-N
SMILES: CC1=NC=C(N1)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (4)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (4)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (4)
  • (1)